Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Odf3l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003497916.2 | XP_003497964.1 | 3 - 4 | 301 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |