Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcne2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003495559.1 | XP_003495607.1 | 1 | 323 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Ion Channel 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |