Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptgir | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003510523.1 | XP_003510571.1 | 1 - 2 | 405 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Apoptosis G-protein coupled receptor Probe spans exons |