Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zscan29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007621739.1 | XP_007619929.1 | 4 - 5 | 1734 | 102 | |
XM_007621745.1 | XP_007619935.1 | 4 - 5 | 1700 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Probe spans exons |