Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Marveld3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003501077.1 | XP_003501125.1 | 1 - 2 | 60 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |