Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Marveld3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003501077.1 | XP_003501125.1 | 3 - 4 | 300 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |