Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nat8l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003504123.1 | XP_003504171.1 | 1 - 2 | 97 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |