Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ahnak2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007633200.1 | XP_007631390.1 | 3 - 4 | 290 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Probe spans exons |