Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chac2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003502727.2 | XP_003502775.1 | 1 - 2 | 274 | 71 | |
GenBank mRNA | JP050944.1 | - | 1 - 2 | 180 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Transporter Recommended Assay Probe spans exons |