Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vnn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003510901.2 | XP_003510949.1 | 6 - 7 | 1598 | 64 | |
GenBank mRNA | JI863792.1 | - | 6 - 7 | 1598 | 64 | |
JP063718.1 | - | 6 - 7 | 1595 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |