Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Leprel2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007646914.1 | XP_007645104.1 | 4 - 5 | 524 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Probe spans exons |