Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptgfrn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007641376.1 | XP_007639566.1 | 8 - 9 | 2497 | 76 | |
GenBank mRNA | JI878885.1 | - | 6 - 7 | 1997 | 76 | |
JP038778.1 | - | 6 - 7 | 1997 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Probe spans exons |