Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrn1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003504286.1 | XP_003504334.1 | 2 - 3 | 192 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |