Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007647375.1 | XP_007645565.1 | 5 - 6 | 792 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Protease Probe spans exons |