Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcee | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003505671.2 | XP_003505719.1 | 2 - 3 | 474 | 79 | |
XM_007646150.1 | XP_007644340.1 | 3 - 4 | 646 | 79 | ||
GenBank mRNA | JI871444.1 | - | 2 - 3 | 482 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |