Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ism2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007640406.1 | XP_007638596.1 | 3 - 4 | 738 | 81 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Probe spans exons |