Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hoxd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007626360.1 | XP_007624550.1 | 2 - 3 | 412 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Probe spans exons |