Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007644832.1 | XP_007643022.1 | 2 - 3 | 330 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |