Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Samd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007652775.1 | XP_007650965.1 | 4 - 5 | 850 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |