Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Enpp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003509853.1 | XP_003509901.1 | 2 - 3 | 1011 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Probe spans exons |