Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syngr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003512192.2 | XP_003512240.1 | 4 - 5 | 650 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |