Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgol1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003500455.2 | XP_003500503.1 | 4 - 5 | 561 | 82 | |
GenBank mRNA | JP052645.1 | - | 1 - 2 | 49 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |