Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cmpk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003501343.2 | XP_003501391.1 | 2 - 3 | 108 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |