Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp64 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003514232.2 | XP_003514280.1 | 1 - 2 | 436 | 62 | |
XM_007654418.1 | XP_007652608.1 | 1 - 2 | 536 | 62 | ||
GenBank mRNA | JI885451.1 | - | 1 - 2 | 373 | 62 | |
JP039274.1 | - | 1 - 2 | 373 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |