Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tekt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003501571.1 | XP_003501619.1 | 1 - 2 | 528 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |