Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Las1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003513914.2 | XP_003513962.1 | 9 - 10 | 1308 | 64 | |
XM_007654156.1 | XP_007652346.1 | 10 - 11 | 1359 | 64 | ||
XM_007654157.1 | XP_007652347.1 | 10 - 11 | 1359 | 64 | ||
GenBank mRNA | JI893876.1 | - | 9 - 10 | 1309 | 64 | |
JP051388.1 | - | 9 - 10 | 1309 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |