Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mettl11b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003505528.2 | XP_003505576.1 | 2 - 3 | 328 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |