Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Laptm4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003504073.2 | XP_003504121.1 | 5 - 6 | 448 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Transporter 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |