Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pllp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007644212.1 | XP_007642402.1 | 3 - 4 | 632 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Ion Channel Probe spans exons |