Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galnt8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003496085.1 | XP_003496133.1 | 6 - 7 | 1103 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |