Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fev | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003507620.2 | XP_003507668.2 | 3 - 4 | 391 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |