Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ebln2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007643820.1 | XP_007642010.1 | 1 - 2 | 127 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Probe spans exons |