Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007644151.1 | XP_007642341.1 | 11 - 12 | 1697 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Protein Kinase Probe spans exons |