Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003506269.2 | XP_003506317.1 | 3 - 4 | 356 | 126 | |
GenBank mRNA | JP057628.1 | - | 3 - 4 | 344 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Probe spans exons |