Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klrg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007643833.1 | XP_007642023.1 | 1 - 2 | 112 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |