Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003508297.2 | XP_003508345.1 | 2 - 3 | 657 | 81 | |
GenBank mRNA | JI894144.1 | - | 1 - 2 | 159 | 81 | |
JP049783.1 | - | 1 - 2 | 159 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |