Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nsun4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007611028.1 | XP_007609218.1 | 4 - 5 | 804 | 87 | |
GenBank mRNA | JP041943.1 | - | 4 - 5 | 771 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |