Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsen2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007647912.1 | XP_007646102.1 | 11 - 12 | 6105 | 74 | |
XM_007647913.1 | XP_007646103.1 | 11 - 12 | 5260 | 74 | ||
XM_007647914.1 | XP_007646104.1 | 11 - 12 | 4523 | 74 | ||
XM_007647915.1 | XP_007646105.1 | 11 - 12 | 4560 | 74 | ||
XM_007647916.1 | XP_007646106.1 | 10 - 11 | 6578 | 74 | ||
XM_007647917.1 | XP_007646107.1 | 11 - 12 | 4444 | 74 | ||
XM_007647918.1 | XP_007646108.1 | 12 - 13 | 2610 | 74 | ||
XM_007647919.1 | XP_007646109.1 | 11 - 12 | 2531 | 74 | ||
XM_007647920.1 | XP_007646110.1 | 10 - 11 | 2269 | 74 | ||
XM_007647921.1 | XP_007646111.1 | 10 - 11 | 1278 | 74 | ||
XM_007647922.1 | XP_007646112.1 | 10 - 11 | 1522 | 74 | ||
XM_007647923.1 | XP_007646113.1 | 9 - 10 | 6301 | 74 | ||
XM_007647924.1 | XP_007646114.1 | 9 - 10 | 1218 | 74 | ||
XM_007647925.1 | XP_007646115.1 | 9 - 10 | 1118 | 74 | ||
GenBank mRNA | JI895875.1 | - | 5 - 6 | 690 | 74 | |
JP060073.1 | - | 5 - 6 | 690 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |