Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppm1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003503263.2 | XP_003503311.1 | 2 - 3 | 725 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Protein Phosphatase Probe spans exons |