Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sult1c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003514252.1 | XP_003514300.1 | 5 - 6 | 610 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Probe spans exons |