Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003509681.2 | XP_003509729.1 | 5 - 6 | 568 | 73 | |
GenBank mRNA | JI896129.1 | - | 5 - 6 | 552 | 73 | |
JP056872.1 | - | 5 - 6 | 552 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Probe spans exons |