Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhdc9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003506858.2 | XP_003506906.2 | 3 - 4 | 1056 | 65 | |
XM_007647391.1 | XP_007645581.1 | 3 - 4 | 1134 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |