Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtfp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003509012.2 | XP_003509060.1 | 2 - 3 | 1449 | 76 | |
XM_007649642.1 | XP_007647832.1 | 3 - 4 | 806 | 76 | ||
XM_007649643.1 | XP_007647833.1 | 3 - 4 | 1029 | 76 | ||
XM_007649644.1 | XP_007647834.1 | 3 - 4 | 821 | 76 | ||
XM_007649645.1 | XP_007647835.1 | 3 - 4 | 719 | 76 | ||
XM_007649646.1 | XP_007647836.1 | 3 - 4 | 942 | 76 | ||
XM_007649647.1 | XP_007647837.1 | 3 - 4 | 734 | 76 |