Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam89a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003496834.1 | XP_003496882.1 | 3 - 4 | 433 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |