Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003509509.2 | XP_003509557.1 | 13 - 14 | 1852 | 60 | |
XM_007650093.1 | XP_007648283.1 | 13 - 14 | 1533 | 60 | ||
XM_007650094.1 | XP_007648284.1 | 13 - 14 | 1852 | 60 | ||
XM_007650095.1 | XP_007648285.1 | 13 - 14 | 1852 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |