Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cc2d2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003497772.1 | XP_003497820.1 | 3 - 4 | 390 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |