Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hebp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003501269.2 | XP_003501317.1 | 2 - 3 | 332 | 74 | |
GenBank mRNA | JI874658.1 | - | 2 - 3 | 324 | 74 | |
JP044806.1 | - | 2 - 3 | 325 | 74 |