Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003512317.1 | XP_003512365.1 | 9 - 10 | 1342 | 101 | |
XM_003512318.1 | XP_003512366.1 | 8 - 9 | 1216 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |