Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stxbp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003514867.2 | XP_003514915.1 | 15 - 16 | 1454 | 65 | |
XM_007654905.1 | XP_007653095.1 | 15 - 16 | 1455 | 65 | ||
GenBank mRNA | JI894043.1 | - | 15 - 16 | 1447 | 65 | |
JP062973.1 | - | 15 - 16 | 1447 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |