Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tab2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498946.2 | XP_003498994.1 | 3 - 4 | 1827 | 63 | |
GenBank mRNA | JI883753.1 | - | 2 - 3 | 1724 | 63 | |
JP062381.1 | - | 2 - 3 | 1724 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |